APO455829145 Apollo Scientific Ltd

Ethyl 1,3-dimethyl-4-(4-methoxyphenoxy)-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD00067964
  • Cas Number : 174842-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Synonym 1,3-Dimethyl-5-(ethoxycarbonyl)-4-(4-methoxyphenoxy)-1H-pyrazolo[3,4-b]pyridine, 4-{[1,3-Dimethyl-5-(ethoxycarbonyl)-1H-pyrazolo[3,4-b]pyridin-4-yl]oxy}anisole
Storage temp. Ambient temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21401

Safety Information

signalword Warning
pcodes P280;P260;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21401
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart