APO455829149 Apollo Scientific Ltd

Butyl 2-chloroacetate

  • MDL number MFCD00018925
  • Cas Number : 590-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.07
Flash Point 36
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 164
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21493

Safety Information

signalword Danger
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21493
pictograms GHS02;GHS05
hstatements H226;H314
Availability details for
Quantity
  • Availability
  • Add to Cart