APO455829153 Apollo Scientific Ltd

(4,5-Dichloro-1H-imidazol-1-yl)methyl (4-chlorophenyl)carbamate

  • MDL number MFCD00114451
  • Cas Number : 650592-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-({[(4-Chlorophenyl)carbamoyl]oxy}methyl)-4,5-dichloro-1H-imidazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21567

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21567
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart