APO455829158 Apollo Scientific Ltd

5-(Thien-2-yl)tetrahydrothiophen-3-one

  • MDL number MFCD00052287
  • Cas Number : 108372-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-58
Synonym 2,3-Dihydro-2,2'-bithiophen-4(5H)-one
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21625

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=21625
Availability details for
Quantity
  • Availability
  • Add to Cart