APO455829159 Apollo Scientific Ltd

4,4-Dimethyl-1,3-oxazolidine-2-thione

  • MDL number MFCD00052156
  • Cas Number : 54013-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-128
Synonym 4,4-Dimethyl-2-thioxo-1,3-oxazolidine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21626

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21626
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart