APO455829160 Apollo Scientific Ltd

3-Phenoxythiophene-2-carboxaldehyde

  • MDL number MFCD00178748
  • Cas Number : 132706-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Formyl-3-phenoxythiophene, 3-Phenoxy-2-thenaldehyde, [(2-Formylthien-3-yl)oxy]benzene
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 184/15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21634

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21634
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart