APO455829174 Apollo Scientific Ltd

1,4-Dimethyl-2-(hydroxymethyl)piperazine

  • MDL number MFCD00174342
  • Cas Number : 14675-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1,4-Dimethylpiperazin-2-yl)methanol
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 114-115/ 15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21779

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21779
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart