APO455829180 Apollo Scientific Ltd

6,7-Dimethoxy-1H-quinazolin-4-one

  • MDL number MFCD07776135
  • Cas Number : 13794-72-4
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 298
Iupac Name 6,7-dimethoxy-4(1H)-quinazolinone
Inchi Code 1S/C10H10N2O3/c1-14-8-3-6-7(4-9(8)15-2)11-5-12-10(6)13/h3-5H,1-2H3,(H,11,12,13)
Inchi Key DMSRMHGCZUXCMJ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 298
Product Link https://store.apolloscientific.co.uk/product/67-dimethoxy-1h-quinazolin-4-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/67-dimethoxy-1h-quinazolin-4-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart