APO455829185 Apollo Scientific Ltd

Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate

  • MDL number MFCD00830964
  • Cas Number : 17740-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Benzyl-2,5-bis(ethoxycarbonyl)pyrrolidine, {[2,5-Bis(ethoxycarbonyl)pyrrolidin-1-yl]methyl}benzene
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21905

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21905
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart