APO455829187 Apollo Scientific Ltd

5-Bromo-3-(2-hydroxyethyl)-1H-indole

  • MDL number MFCD00130167
  • Cas Number : 32774-29-1
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Synonym 5-Bromotryptophol
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-2-hydroxyethyl-1h-indole

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-2-hydroxyethyl-1h-indole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart