APO455829191 Apollo Scientific Ltd

2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)nicotinamide

  • MDL number MFCD01566566
  • Cas Number : 680211-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-46
Synonym 2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)pyridine-3-carboxamide, 2-Chloro-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)pyridine-3-carboxamide
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21935

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21935
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart