APO455829192 Apollo Scientific Ltd

3,5-Dimethyl-N-(2-{[(fur-2-yl)methyl]thio}ethyl)isoxazole-4-sulphonamide

  • MDL number MFCD01570599
  • Cas Number : 680211-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,5-Dimethyl-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)-1,2-oxazole-4-sulphonamide, 2-{[(2-{[(3,5-Dimethyl-1,2-oxazol-4-yl)sulphonyl]amino}ethyl)sulphanyl]methyl}furan
Storage temp. Ambient temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21939

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21939
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart