APO455829196 Apollo Scientific Ltd

4-Methoxybenzyl chloride

  • MDL number MFCD00000915
  • Cas Number : 824-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -1
Synonym 1-(Chloromethyl)-4-methoxybenzene, 4-(Chloromethyl)anisole
Density 1.15
Flash Point 109
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 117-118/14mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21961

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21961
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart