APO455829197 Apollo Scientific Ltd

2-Chloro-4-nitro-1H-imidazole

  • MDL number MFCD03419295
  • Cas Number : 57531-37-0
  • Molecular Weight : 147.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 214-216
Iupac Name 2-chloro-4-nitro-1H-imidazole
Inchi Code 1S/C3H2ClN3O2/c4-3-5-1-2(6-3)7(8)9/h1H,(H,5,6)
Inchi Key BOJZBRDIZUHTCE-UHFFFAOYSA-N

Description

Synonym 2-Chloro-5-nitro-1H-imidazole
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 214-216
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-nitro-1h-imidazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-nitro-1h-imidazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart