APO455829198 Apollo Scientific Ltd

4-Chloro-N-ethyl-6-(1-phenyl-1H-indol-3-yl)-1,3,5-triazin-2-amine

  • MDL number MFCD00123281
  • Cas Number : 500587-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[4-Chloro-6-(ethylamino)-1,3,5-triazin-2-yl]-1-phenyl-1H-indole, {3-[4-Chloro-6-(ethylamino)-1,3,5-triazin-2-yl]-1H-indol-1-yl}benzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28196

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28196
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart