APO455829205 Apollo Scientific Ltd

3,4-Dichloro-N-[(diphenylphosphoryl)(fur-2-yl)methyl]aniline

  • MDL number MFCD00123332
  • Cas Number : 680212-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-{[(3,4-Dichlorophenyl)amino](diphenylphosphoryl)methyl}furan
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=21998

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=21998
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart