APO455829207 Apollo Scientific Ltd

N-Benzyl-2-nitro-3,4,5,6-tetrachloroaniline

  • MDL number MFCD00114878
  • Cas Number : 69035-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Benzylamino)-3,4,5,6-tetrachloronitrobenzene, N-(2-Nitro-3,4,5,6-tetrachlorophenyl)benzylamine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22029

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22029
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart