APO455829217 Apollo Scientific Ltd

5-Phenyl-1,3,4-oxadiazole-2-carbonyl chloride

  • MDL number MFCD03086204
  • Cas Number : 98591-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-90
Synonym 2-(Chlorocarbonyl)-5-phenyl-1,3,4-oxadiazole, 2-(Chloroformyl)-5-phenyl-1,3,4-oxadiazole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22099

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22099
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart