APO455829227 Apollo Scientific Ltd

Methyl 1H-benzimidazole-5-carboxylate

  • MDL number MFCD03407310
  • Cas Number : 26663-77-4
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-1h-benzimidazole-5-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-1h-benzimidazole-5-carboxylate
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart