APO455829231 Apollo Scientific Ltd

3-Amino-1,2-benzisoxazole

  • MDL number MFCD03407353
  • Cas Number : 36216-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Synonym 1,2-Benzisoxazol-3-amine, Benzo[d]isoxazol-3-amine
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22148

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22148
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart