APO455829234 Apollo Scientific Ltd

4-Methyl-1,3-thiazole-2-carboxylic acid

  • MDL number MFCD07377486
  • Cas Number : 14542-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Synonym 2-Carboxy-4-methyl-1,3-thiazole
Purity 96
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Ship on Ice
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38675

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38675
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart