APO455829237 Apollo Scientific Ltd

2,2-Dimethylchroman-6-sulphonyl chloride

  • MDL number MFCD03407324
  • Cas Number : 131880-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Synonym 3,4-Dihydro-2,2-dimethyl-2H-chromene-6-sulphonyl chloride, 3,4-Dihydro-2,2-dimethyl-2H-1-benzopyran-6-sulphonyl chloride
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22157

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22157
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart