APO455829240 Apollo Scientific Ltd

5-Methyl-1,3-thiazole-2-carboxylic acid

  • MDL number MFCD08060736
  • Cas Number : 61291-21-2
  • Molecular Weight : 143.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-1,3-thiazole-2-carboxylic acid
Inchi Code 1S/C5H5NO2S/c1-3-2-6-4(9-3)5(7)8/h2H,1H3,(H,7,8)
Inchi Key UYDGNRLMYZMDOG-UHFFFAOYSA-N

Description

Synonym 2-Carboxy-5-methyl-1,3-thiazole
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methyl-13-thiazole-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-13-thiazole-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart