APO455829242 Apollo Scientific Ltd

6-Bromoimidazo[1,2-a]pyrazine-2-carboxylic acid

  • MDL number MFCD09414724
  • Cas Number : 1000018-56-3
  • Molecular Weight : 242.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Carboxy-6-bromoimidazo[1,2-a]pyrazine
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromoimidazo12-apyrazine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromoimidazo12-apyrazine-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart