APO455829243 Apollo Scientific Ltd

3-(2-Methylpyrimidin-4-yl)benzenesulphonyl chloride

  • MDL number MFCD03659718
  • Cas Number : 465514-07-2
  • Molecular Weight : 268.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 174-175
Iupac Name 3-(2-methyl-4-pyrimidinyl)benzenesulfonyl chloride
Inchi Code 1S/C11H9ClN2O2S/c1-8-13-6-5-11(14-8)9-3-2-4-10(7-9)17(12,15)16/h2-7H,1H3
Inchi Key ZLLPUDLQNCNHFM-UHFFFAOYSA-N

Description

Synonym 4-[3-(Chlorosulphonyl)phenyl]-2-methylpyrimidine, 3-(2-Methyl-1,3-diazin-4-yl)benzenesulphonyl chloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 174-175
Product Link https://store.apolloscientific.co.uk/product/3-2-methylpyrimidin-4-ylbenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-2-methylpyrimidin-4-ylbenzenesulphonyl-chloride
pictograms GHS05
hstatements H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart