APO455829245 Apollo Scientific Ltd

2-Cyano-5-methylbenzenesulphonyl chloride

  • MDL number MFCD03659720
  • Cas Number : 197960-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67
Synonym 2-(Chlorosulphonyl)-4-methylbenzonitrile, 3-(Chlorosulphonyl)-4-cyanotoluene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22184

Safety Information

signalword Danger
pcodes P260;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22184
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart