APO455829251 Apollo Scientific Ltd

2-(Bromoacetyl)-3-methylbenzo[b]thiophene

  • MDL number MFCD00173793
  • Cas Number : 118337-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-(3-methylbenzo[b]thiophen-2-yl)ethan-1-one
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22213

Safety Information

signalword Danger
pcodes P280;P301+P330+P331;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22213
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart