APO455829252 Apollo Scientific Ltd

1-[(4-Methylphenyl)thio]acetone

  • MDL number MFCD00052713
  • Cas Number : 1200-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 96-98/0.1mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22215

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22215
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart