APO455829255 Apollo Scientific Ltd

2-(Hydroxymethyl)-5-methyl-1,3-thiazole

  • MDL number MFCD08060774
  • Cas Number : 202932-04-5
  • Molecular Weight : 129.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-methyl-1,3-thiazol-2-yl)methanol
Inchi Code 1S/C5H7NOS/c1-4-2-6-5(3-7)8-4/h2,7H,3H2,1H3
Inchi Key BWKAOFJDCPNWEC-UHFFFAOYSA-N

Description

Synonym (5-Methyl-1,3-thiazol-2-yl)methanol
Storage temp. Store at 2-8
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxymethyl-5-methyl-13-thiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxymethyl-5-methyl-13-thiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart