APO455829259 Apollo Scientific Ltd

4-[(Methylsulphonyl)oxy]phenylacetic acid

  • MDL number MFCD09800580
  • Cas Number : 64369-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Carboxymethyl)-4-[(methylsulphonyl)oxy]benzene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40606

Safety Information

signalword Danger
pcodes P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40606
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart