APO455829264 Apollo Scientific Ltd

2-Methylpyridine-4-boronic acid

  • MDL number MFCD03095364
  • Cas Number : 579476-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-123
Synonym 4-Borono-2-methylpyridine
Storage temp. Store at -20
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-methylpyridine-4-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylpyridine-4-boronic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart