APO455829268 Apollo Scientific Ltd

5-Aminoindan-1-one

  • MDL number MFCD00099466
  • Cas Number : 3470-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-186
Synonym 5-Amino-2,3-dihydro-1H-inden-1-one, 5-Amino-2,3-dihydro-1-oxo-1H-indene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-aminoindan-1-one

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-aminoindan-1-one
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart