APO455829271 Apollo Scientific Ltd

Quinoline-5-boronic acid

  • MDL number MFCD03095058
  • Cas Number : 355386-94-6
  • Molecular Weight : 172.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-quinolinylboronic acid
Inchi Code 1S/C9H8BNO2/c12-10(13)8-4-1-5-9-7(8)3-2-6-11-9/h1-6,12-13H
Inchi Key NWIJBOCPTGHGIK-UHFFFAOYSA-N

Description

Synonym 5-Boronoquinoline
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/quinoline-5-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/quinoline-5-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart