APO455829272 Apollo Scientific Ltd

Quinoline-6-boronic acid

  • MDL number MFCD03093100
  • Cas Number : 376581-24-7
  • Molecular Weight : 172.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 106-109
Iupac Name 6-quinolinylboronic acid
Inchi Code 1S/C9H8BNO2/c12-10(13)8-3-4-9-7(6-8)2-1-5-11-9/h1-6,12-13H
Inchi Key JLOLSBLXNMVKGY-UHFFFAOYSA-N

Description

Synonym 6-Boronoquinoline, 6-Borono-1-azanaphthalene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 106-109
Product Link https://store.apolloscientific.co.uk/product/quinoline-6-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/quinoline-6-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart