APO455829276 Apollo Scientific Ltd

1-Oxaspiro[2.5]octane-2-carbonitrile

  • MDL number MFCD00101211
  • Cas Number : 36929-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Cyano-1-oxaspiro[2.5]octane
Density 1.0275
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 84-85/5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22450

Safety Information

signalword Danger
pcodes P261;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22450
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart