APO455829279 Apollo Scientific Ltd

4-Amino-N,N-dimethylbenzamide

  • MDL number MFCD00523648
  • Cas Number : 6331-71-1
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 151-154
Iupac Name 4-amino-N,N-dimethylbenzamide
Inchi Code 1S/C9H12N2O/c1-11(2)9(12)7-3-5-8(10)6-4-7/h3-6H,10H2,1-2H3
Inchi Key QEPGWLBMAAEBCP-UHFFFAOYSA-N

Description

Synonym 4-(Dimethylcarbamoyl)aniline
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 151-154
Product Link https://store.apolloscientific.co.uk/product/4-amino-nn-dimethylbenzamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-nn-dimethylbenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart