APO455829284 Apollo Scientific Ltd

2-Bromo-4-(hydroxymethyl)phenol

  • MDL number MFCD00016869
  • Cas Number : 29922-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-132
Synonym 3-Bromo-4-hydroxybenzyl alcohol, (3-Bromo-4-hydroxyphenyl)methanol
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-hydroxymethylphenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-hydroxymethylphenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart