APO455829285 Apollo Scientific Ltd

5-Chloro-2-methoxy-4-nitroaniline

  • MDL number MFCD00126494
  • Cas Number : 6259-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-133
Synonym 2-Amino-4-chloro-5-nitroanisole, 5-Chloro-4-nitro-o-anisidine, 4-Amino-2-chloro-5-methoxynitrobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=22641

Safety Information

signalword Warning
pcodes P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=22641
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart