APO455829290 Apollo Scientific Ltd

3-Chloro-1,2-benzisothiazole

  • MDL number MFCD00673254
  • Cas Number : 7716-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38-39
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 80-86/0.75mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37965

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37965
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart