APO455829301 Apollo Scientific Ltd

Methyl 3-(bromomethyl)-4-cyanobenzoate

  • MDL number MFCD09839300
  • Cas Number : 908562-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Cyano-5-(methoxycarbonyl)benzyl bromide, 2-(Bromomethyl)-4-(methoxycarbonyl)benzonitrile
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40330

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40330
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart