APO455829305 Apollo Scientific Ltd

{4-[(tert-Butoxycarbonyl)amino]tetrahydro-2H-pyran-4-yl}acetic acid

  • MDL number MFCD09264196
  • Cas Number : 946682-30-0
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[(tert-Butoxycarbonyl)amino]-4-(carboxymethyl)tetrahydro-2H-pyran, 4-Amino-4-(carboxymethyl)tetrahydro-2H-pyran, N-BOC protected
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-tert-butoxycarbonylaminotetrahydro-2h-pyran-4-ylacetic-acid

Safety Information

signalword Warning
pcodes P280;P304+P340;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-tert-butoxycarbonylaminotetrahydro-2h-pyran-4-ylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart