APO455829307 Apollo Scientific Ltd

[4-({{(9H-Fluoren-9-yl)methoxy]carbonyl}amino)tetrahydro-2H-pyran-4-yl]acetic acid

  • MDL number MFCD09264206
  • Cas Number : 946716-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (9H-Fluoren-9-yl)methyl [4-(carboxymethyl)tetrahydro-2H-pyran-4-yl]carbamate, 4-Amino-4-(carboxymethyl)tetrahydro-2H-pyran, N-FMOC protected
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42531

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42531
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart