APO455829309 Apollo Scientific Ltd

2-Amino-4-(4-methoxyphenyl)-1,3-thiazole

  • MDL number MFCD00170663
  • Cas Number : 2104-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-209
Synonym 4-(4-Methoxyphenyl)-1,3-thiazol-2-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-4-methoxyphenyl-13-thiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-4-methoxyphenyl-13-thiazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart