APO455829312 Apollo Scientific Ltd

5-Amino-1,2-dihydroacenaphthylene

  • MDL number MFCD00029136
  • Cas Number : 4657-93-6
  • Molecular Weight : 169.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-108
Synonym 1,2-Dihydroacenaphthylen-5-amine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-12-dihydroacenaphthylene

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-12-dihydroacenaphthylene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart