APO455829317 Apollo Scientific Ltd

5-Bromonicotinoyl chloride

  • MDL number MFCD00173853
  • Cas Number : 39620-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-74
Synonym 5-Bromopyridine-3-carbonyl chloride, 3-Bromo-5-(chlorocarbonyl)pyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromonicotinoyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromonicotinoyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart