APO455829324 Apollo Scientific Ltd

2-Methoxybenzophenone

  • MDL number MFCD00017163
  • Cas Number : 2553-04-0
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 36-38
Iupac Name (2-methoxyphenyl)(phenyl)methanone
Inchi Code 1S/C14H12O2/c1-16-13-10-6-5-9-12(13)14(15)11-7-3-2-4-8-11/h2-10H,1H3
Inchi Key CSUUDNFYSFENAE-UHFFFAOYSA-N

Description

Synonym (2-Methoxyphenyl)(phenyl)methanone, 2-Benzoylanisole
Flash Point 149-150/1mm
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 36-38
Product Link https://store.apolloscientific.co.uk/product/2-methoxybenzophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxybenzophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart