APO455829330 Apollo Scientific Ltd

2-(4-Chlorophenoxy)-2-methylpropanoyl chloride

  • MDL number MFCD00128244
  • Cas Number : 5542-60-9
  • Molecular Weight : 233.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-chlorophenoxy)-2-methylpropanoyl chloride
Inchi Code 1S/C10H10Cl2O2/c1-10(2,9(12)13)14-8-5-3-7(11)4-6-8/h3-6H,1-2H3
Inchi Key OODRWLGKUBMFLZ-UHFFFAOYSA-N

Description

Synonym 2-(4-Chlorophenoxy)-2,2-dimethylacetyl chloride
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-4-chlorophenoxy-2-methylpropanoyl-chloride

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-4-chlorophenoxy-2-methylpropanoyl-chloride
pictograms GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart