APO455829331 Apollo Scientific Ltd

2,6-Dimethyl-4-hydroxybenzonitrile

  • MDL number MFCD00238591
  • Cas Number : 58537-99-8
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 169-173
Iupac Name 4-hydroxy-2,6-dimethylbenzonitrile
Inchi Code 1S/C9H9NO/c1-6-3-8(11)4-7(2)9(6)5-10/h3-4,11H,1-2H3
Inchi Key KZEJTKHRBQWACL-UHFFFAOYSA-N

Description

Synonym 4-Cyano-3,5-dimethylphenol, 2-Cyano-5-hydroxy-m-xylene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 169-173
Product Link https://store.apolloscientific.co.uk/product/26-dimethyl-4-hydroxybenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dimethyl-4-hydroxybenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart