APO455829334 Apollo Scientific Ltd

(2,3-Dihydrobenzo[b]furan-5-yl)acetic acid

  • MDL number MFCD01941301
  • Cas Number : 69999-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-107
Synonym 5-(Carboxymethyl)-2,3-dihydrobenzo[b]furan, 2-(2,3-Dihydro-1-benzofuran-5-yl)ethanoic acid
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16024

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16024
pictograms GHS05;GHS07
hstatements H302;H312;H332;H317;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart