APO455829339 Apollo Scientific Ltd

N1-(2,3-Dichlorophenyl)-2-chloroacetamide

  • MDL number MFCD00028173
  • Cas Number : 33560-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-106
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23215

Safety Information

signalword Warning
pcodes P260;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23215
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart